Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74564

Supelco

Trifluoroacetic acid

analytical standard

Synonym(s):

TFA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COOH
CAS Number:
Molecular Weight:
114.02
Beilstein:
742035
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.9 (vs air)

vapor pressure

97.5 mmHg ( 20 °C)

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.05% water

refractive index

n20/D 1.3 (lit.)

bp

72.4 °C (lit.)

mp

−15.4 °C (lit.)

solubility

H2O: soluble

density

1.489 g/mL at 20 °C (lit.)

application(s)

environmental

format

neat

SMILES string

OC(C(F)(F)F)=O

InChI

1S/C2HF3O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

DTQVDTLACAAQTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trifluoroacetic acid is a phytotoxic compound belonging to the class of perfluorinated acids (PFAs). It is an environmental contaminant, which contributes to the organo-fluorine burden in surface and ground waters.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Trifluoroacetic acid may be used as a modifier in high performance liquid chromatography mobile phase for protein and peptide separation due to its properties like chromatographic selectivity, high volatility and low ultraviolet light cut-off.
Trifluoroacetic acid may be used as an analytical reference standard for the determination of the analyte in water matrices using high-performance liquid chromatography coupled to tandem mass spectrometry.
Trifluoroacetic acid may be used as a modifier in high performance liquid chromatography mobile phase for protein and peptide separation due to its properties like chromatographic selectivity, high volatility and low ultraviolet light cut-off.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

>212.0 °F - Pensky-Martens closed cup

Flash Point(C)

> 100 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced sensitivity for peptide mapping with electrospray liquid chromatography-mass spectrometry in the presence of signal suppression due to trifluoroacetic acid-containing mobile phases
Apffel A, et al.
Journal of Chromatography A, 712(1), 177-190 (1995)
Analysis of trifluoroacetic acid and other short-chain perfluorinated acids (C2--C4) in precipitation by liquid chromatography-tandem mass spectrometry: Comparison to patterns of long-chain perfluorinated acids (C5--C18)
Taniyasu S, et al.
Analytica Chimica Acta, 619(2), 221-230 (2008)
Yukiko Enomoto-Rogers et al.
Carbohydrate polymers, 92(2), 1827-1834 (2013-02-13)
Fully substituted glucomannan triacetate (GMTAc) (degree of substitution (DS)=3.0) was prepared from konjac glucomannan (KGM) treated with acetic acid and trifluoroacetic anhydride (TFAA). The peaks in the (1)H- and (13)C NMR spectra of GMTAc were assigned in detail based on
Caroline Le Maréchal et al.
Veterinary research, 42, 35-35 (2011-02-18)
Staphylococcus aureus is a major cause of mastitis in ruminants. In ewe mastitis, symptoms range from subclinical to gangrenous mastitis. S. aureus factors or host-factors contributing to the different outcomes are not completely elucidated. In this study, experimental mastitis was
Wolfgang Klein et al.
PloS one, 10(3), e0120886-e0120886 (2015-03-26)
The cyclodepsipeptide cotransin was described to inhibit the biosynthesis of a small subset of proteins by a signal sequence-discriminatory mechanism at the Sec61 protein-conducting channel. However, it was not clear how selective cotransin is, i.e. how many proteins are sensitive.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service