Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43492

Supelco

Sodium Standard for IC

TraceCERT®, 1000 mg/L Na+ in water (nominal concentration), Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

1000 mg/L Na+ in water (nominal concentration)

technique(s)

ion chromatography: suitable

application(s)

environmental
food and beverages

format

single component solution

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

It is intended to be used during the calibration of ion chromatography-based analytical techniques.

Features and Benefits

  • Produced under ISO/IEC 17025 and ISO 17034 accreditation
  • Traceable to SI unit of mole through internationally accepted institutions such as NIST or BAM
  • Accompanied by a comprehensive Certificate of Analysis (CoA) (up to four pages) with data on stability, density, homogeneity, the accuracy of concentration, uncertainty, and traceability
  • Gravimetrically prepared using high-quality qNMR-characterized starting materials and qualified precision balances to ensure minimal uncertainty
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Available in an HDPE bottle, sealed in an aluminum bag

Linkage

Visit the IC Portal to learn more

Analysis Note

1000 mg/L Na+ in water, prepared with high purity NaNO3 and water

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicole A P van Gestel et al.
Journal of materials science. Materials in medicine, 30(8), 94-94 (2019-08-16)
Clinically, S53P4 bioactive glass (BAG) has shown very promising results in bone infection treatment, but it is also known to degrade very slowly in vivo. To evaluate which mechanisms (cellular or dissolution) can play a role in the degradation of

Articles

Metrosep C Supp 2 column and acidic binary IC eluent accurately determine ammonium in acidic solutions, meeting pharmacopeia regulations.

Metrosep C Supp 2 column and acidic binary IC eluent accurately determine ammonium in acidic solutions, meeting pharmacopeia regulations.

Metrosep C Supp 2 column and acidic binary IC eluent accurately determine ammonium in acidic solutions, meeting pharmacopeia regulations.

Metrosep C Supp 2 column and acidic binary IC eluent accurately determine ammonium in acidic solutions, meeting pharmacopeia regulations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service