Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

P8650

Sigma-Aldrich

Prednisolone 21-acetate

≥97%

Synonym(s):

1,4-Pregnadiene-11β,17α,21-triol-3,20-dione 21-acetate, 11β,17α,21-Trihydroxy-1,4-pregnadiene-3,20-dione 21-acetate, 21-Acetoxy-1,4-pregnadiene-11β,17α-diol-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H30O6
CAS Number:
Molecular Weight:
402.48
Beilstein:
3111798
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

sterility

non-sterile

Assay

≥97%

form

powder

solubility

chloroform: methanol (1:1): 50 mg/mL, clear, colorless

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@@]12CCC3=CC(=O)C=C[C@]3(C)[C@@]1([H])[C@@H](O)C[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(=O)COC(C)=O

InChI

1S/C23H30O6/c1-13(24)29-12-19(27)23(28)9-7-17-16-5-4-14-10-15(25)6-8-21(14,2)20(16)18(26)11-22(17,23)3/h6,8,10,16-18,20,26,28H,4-5,7,9,11-12H2,1-3H3/t16-,17-,18-,20+,21-,22-,23-/m0/s1

InChI key

LRJOMUJRLNCICJ-JZYPGELDSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Namrata Sharma et al.
Cornea, 34(9), 1170-1174 (2015-06-27)
To evaluate the role of anterior segment optical coherence tomography (ASOCT) in the detection and management of Descemet membrane detachment (DMD) in cases of persistent corneal edema after intraocular surgery. An ASOCT-guided new algorithm for the management of such DMDs
Florian Birnbaum et al.
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie, 251(1), 95-103 (2012-05-11)
Postoperative astigmatism following penetrating keratoplasty is a major problem after corneal transplantation. The main goal of new trephination techniques such as femtosecond laser or excimer-laser trephination is to improve refractive and visual outcomes. The femtosecond laser technique makes profiled corneal
Susan M Wakil et al.
Canadian journal of ophthalmology. Journal canadien d'ophtalmologie, 47(4), 339-343 (2012-08-14)
The objective of this study was to present a unique case of unilateral facial nerve palsy as an isolated complication of herpes zoster ophthalmicus. An 82-year-old immunocompetent male presented with a 1-week history of painful left scalp lesions. The diagnosis
Rohit Shetty et al.
Indian journal of ophthalmology, 62(9), 923-926 (2014-11-06)
To study the safety and efficacy of sutureless femtosecond anterior lamellar keratoplasty (FALK) in patients with corneal stromal opacities. Eleven eyes of 11 consecutive patients with corneal stromal opacities involving < 250 μ due to various pathologies were included in
Rohit Shetty et al.
American journal of ophthalmology, 159(3), 419-425 (2014-12-03)
To evaluate the effect of keratoconus cone location on the change in refractive outcomes, corneal aberrations, and biomechanics after combined topography-guided photorefractive keratectomy (PRK) and collagen cross-linking (CXL). Prospective, comparative case series. Topography-guided PRK was performed followed by accelerated CXL

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service