Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0000582

Prednisolone acetate for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Prednisolone 21-acetate, 1,4-Pregnadiene-11β,17α,21-triol-3,20-dione 21-acetate, 11β,17α,21-Trihydroxy-1,4-pregnadiene-3,20-dione 21-acetate, 21-Acetoxy-1,4-pregnadiene-11β,17α-diol-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H30O6
CAS Number:
Molecular Weight:
402.48
Beilstein:
3111798
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

prednisolone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12CCC3=CC(=O)C=C[C@]3(C)[C@@]1([H])[C@@H](O)C[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(=O)COC(C)=O

InChI

1S/C23H30O6/c1-13(24)29-12-19(27)23(28)9-7-17-16-5-4-14-10-15(25)6-8-21(14,2)20(16)18(26)11-22(17,23)3/h6,8,10,16-18,20,26,28H,4-5,7,9,11-12H2,1-3H3/t16-,17-,18-,20+,21-,22-,23-/m0/s1

InChI key

LRJOMUJRLNCICJ-JZYPGELDSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Prednisolone acetate for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcus Ang et al.
American journal of ophthalmology, 157(6), 1163-1169 (2014-03-04)
To compare 3-year endothelial cell loss and graft survival following Descemet stripping automated endothelial keratoplasty (DSAEK) using the EndoGlide (AngioTech, Reading, Pennsylvania, USA/Network Medical Products, North Yorkshire, UK) donor insertion device compared to donor insertion using the Sheets glide technique.
Salima I Hassanaly et al.
American journal of ophthalmology, 158(2), 270-276 (2014-05-23)
To describe outcomes after Boston Type 1 Keratoprosthesis (KPro) surgery in aniridic eyes. Retrospective, interventional case series. University-based tertiary care institution. Twenty-six aniridic eyes of 19 patients who underwent KPro implantation by a single experienced surgeon (M.H.-D.) between October 27
Rohit Shetty et al.
Indian journal of ophthalmology, 62(9), 923-926 (2014-11-06)
To study the safety and efficacy of sutureless femtosecond anterior lamellar keratoplasty (FALK) in patients with corneal stromal opacities. Eleven eyes of 11 consecutive patients with corneal stromal opacities involving < 250 μ due to various pathologies were included in
Oscar A Candia et al.
Experimental eye research, 128, 114-116 (2014-10-12)
We have previously shown that tissue plasminogen activator (tPA) injected in the vitreous of sheep, reduced or prevented the elevation of the intraocular pressure (IOP) normally produced by the instillation of 1% prednisolone. We now report the effect of tPA
Rohit Shetty et al.
American journal of ophthalmology, 159(3), 419-425 (2014-12-03)
To evaluate the effect of keratoconus cone location on the change in refractive outcomes, corneal aberrations, and biomechanics after combined topography-guided photorefractive keratectomy (PRK) and collagen cross-linking (CXL). Prospective, comparative case series. Topography-guided PRK was performed followed by accelerated CXL

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service