Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1537003

USP

Physostigmine salicylate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Eserine salicylate salt, Physostigmine salicylate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21N3O2 · C7H6O3
CAS Number:
Molecular Weight:
413.47
Beilstein:
3900576
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

physostigmine

manufacturer/tradename

USP

mp

181-183 °C

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OC(=O)c1ccccc1O.CNC(=O)Oc2ccc3N(C)[C@H]4N(C)CC[C@@]4(C)c3c2

InChI

1S/C15H21N3O2.C7H6O3/c1-15-7-8-17(3)13(15)18(4)12-6-5-10(9-11(12)15)20-14(19)16-2;8-6-4-2-1-3-5(6)7(9)10/h5-6,9,13H,7-8H2,1-4H3,(H,16,19);1-4,8H,(H,9,10)/t13-,15+;/m1./s1

InChI key

HZOTZTANVBDFOF-PBCQUBLHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Physostigmine salicylate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Physostigmine Salicylate Injection

Biochem/physiol Actions

Acetylcholinesterase inhibitor that crosses the blood-brain barrier and forms a carbamylated enzyme complex with acetyl cholinesterase that degrades slowly.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service