Skip to Content
MilliporeSigma
All Photos(1)

Documents

N0500000

Neostigmine bromide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

3-(N,N-Dimethylcarbamoyloxy)-N,N,N,-trimethylanilinium bromide, Prostigmine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H19BrN2O2
CAS Number:
Molecular Weight:
303.20
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

neostigmine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Br-].CN(C)C(=O)Oc1cccc(c1)[N+](C)(C)C

InChI

1S/C12H19N2O2.BrH/c1-13(2)12(15)16-11-8-6-7-10(9-11)14(3,4)5;/h6-9H,1-5H3;1H/q+1;/p-1

InChI key

LULNWZDBKTWDGK-UHFFFAOYSA-M

Gene Information

human ... ACHE(43)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Neostigmine bromide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Reversible inhibitor of acetylcholinesterase that is similar to eserine, but does not cross the blood-brain barrier.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessica Kåhlin et al.
Experimental physiology, 99(8), 1089-1098 (2014-06-03)
Studies on experimental animals established that the carotid bodies are sensory organs for detecting arterial blood O2 levels and that the ensuing chemosensory reflex is a major regulator of cardiorespiratory functions during hypoxia. However, little information is available on the
Su-Hsun Liu et al.
Clinical toxicology (Philadelphia, Pa.), 53(1), 13-21 (2014-11-21)
Pufferfish-associated tetrodotoxin poisoning occurs occasionally in coastal regions of Asian countries and sporadically elsewhere. While anecdotal reports have suggested neostigmine use, evidence for its clinical effectiveness remains unclear. We sought to determine whether neostigmine was likely to be effective in
Machika Moriya et al.
Masui. The Japanese journal of anesthesiology, 63(4), 418-422 (2014-05-03)
We experienced severe asthmatic crisis during general anesthesia in a 45-year-old man with IgG4-related disease, COPD and athma undergoing removal of submandibular gland. The ventilatiory failure was caused by the stimulation of the operation, sputum, and neostigmine. His serum IgG4
Seham Mustafa et al.
Urology, 84(6), 1549-1549 (2014-12-03)
To examine the effects of neostigmine, an acetylcholinesterase inhibitor that has been used to treat impaired bladder emptying on diabetic rat urinary bladder smooth muscle. Rat urinary detrusor muscle strips were suspended in organ baths containing Krebs' solution for isometric
H S Bawaskar et al.
Tropical doctor, 44(2), 82-85 (2014-02-20)
Between January 2005 and August 2011 141 victims of krait bite poisoning were admitted to the general hospital at Mahad. Clinical signs and symptoms preceding the development of neuroparalysis were analyzed. Fifty-six percent of patients were male. A total of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service