Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1457607

USP

Nateglinide

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Fastic, N-[(trans-4-Isopropylcyclohexyl)carbonyl]-D-phenylalanine, Starlix, Starsis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H27NO3
CAS Number:
Molecular Weight:
317.42
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

nateglinide

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(C)[C@@H]1CC[C@H](CC1)C(=O)N[C@H](Cc2ccccc2)C(O)=O

InChI

1S/C19H27NO3/c1-13(2)15-8-10-16(11-9-15)18(21)20-17(19(22)23)12-14-6-4-3-5-7-14/h3-7,13,15-17H,8-12H2,1-2H3,(H,20,21)(H,22,23)/t15-,16-,17-/m1/s1

InChI key

OELFLUMRDSZNSF-BRWVUGGUSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

Nateglinide is a Kir6.2/SUR1 channel inhibitor and antidiabetic. It is selective for the SUR1 subtype, which is found on pancreatic islet cells. Nateglinide evokes KATP channel-dependent insulin secretion (50-200 μM) in the absence and presence of insulin.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James F McLeod
Clinical pharmacokinetics, 43(2), 97-120 (2004-01-30)
The prevalence and medical and economic impact of type 2 diabetes mellitus is increasing in Western societies. New agents have been developed that act primarily to reduce postprandial glucose excursions, which may be of particular significance now that postprandial glucose
T L Levien et al.
The Annals of pharmacotherapy, 35(11), 1426-1434 (2001-11-29)
To review the pharmacology, pharmacokinetics, dosing guidelines, adverse effects, drug interactions, and clinical efficacy of nateglinide. Primary and review articles regarding nateglinide were identified by MEDLINE search (from 1966 to January 2001); abstracts were identified through the Institute for Scientific
Masato Odawara
Nihon rinsho. Japanese journal of clinical medicine, 61(7), 1230-1237 (2003-07-25)
Patients with type 2 diabetes mellitus are associated with insulin resistance and/or impaired insulin secretion. Previous observations indicate that Japanese patients with type 2 diabetes tend to have impaired insulin response after glycemic load more often than Caucasian counterparts. Recently
George Grunberger
Expert opinion on pharmacotherapy, 12(13), 2097-2106 (2011-08-02)
It has been hypothesized that reducing postprandial glucose spikes would result both in improved overall glycemic control and long-term (especially cardiovascular) outcomes in patients with type 2 diabetes. Nateglinide, a short-acting insulin secretagogue of the meglitinide class of antidiabetic agents
C J Halas
American journal of health-system pharmacy : AJHP : official journal of the American Society of Health-System Pharmacists, 58(13), 1200-1205 (2001-07-14)
The pharmacology, pharmacokinetics, clinical efficacy, adverse effects, interactions, and dosage of nateglinide are reviewed. Nateglinide is an oral hypoglycemic agent approved for use alone or in combination with metformin as an adjunct to diet and exercise for the treatment of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service