MilliporeSigma
  • Clinical pharmacokinetics of nateglinide: a rapidly-absorbed, short-acting insulinotropic agent.
Clinical pharmacokinetics of nateglinide: a rapidly-absorbed, short-acting insulinotropic agent.
Clinical pharmacokinetics (2004-01-30)
James F McLeod
ABSTRACT
The prevalence and medical and economic impact of type 2 diabetes mellitus is increasing in Western societies. New agents have been developed that act primarily to reduce postprandial glucose excursions, which may be of particular significance now that postprandial glucose excursions are known to be correlated with cardiovascular morbidity and mortality. Nateglinide is a phenylalanine derivative that blocks K+ channels in pancreatic beta-cells, facilitating insulin secretion. Nateglinide sensitises beta-cells to ambient glucose, reducing the glucose concentration needed to stimulate insulin secretion. The pharmacokinetics of nateglinide are characterised by rapid absorption and elimination, with good (73%) bioavailability. Nateglinide is more rapidly absorbed when given 0-30 minutes prior to meal ingestion than if given during the meal. Nateglinide is extensively metabolised, primarily by cytochrome P450 2C9, and eliminated primarily by the kidney. Nateglinide pharmacokinetics are linear over the dose range 60-240 mg. No significant pharmacokinetic alterations occur in renally impaired patients, in the elderly, or in mildly hepatically impaired patients. Nateglinide administered prior to meals stimulates rapid, short-lived insulin secretion in a dose-dependent manner, thus decreasing mealtime plasma glucose excursions. Its effects on insulin secretion are synergistic with those of a meal. With increasing nateglinide doses, the risk of hypoglycaemia also increases, but its incidence is low. Even if a meal is missed, and the patient skips the dose of nateglinide (as recommended in the event of a missed meal), the incidence of subsequent hypoglycaemia remains low compared with long-acting agents. The postprandial insulinotropic effects of nateglinide are more rapid than those of repaglinide and more rapid and greater than those of glibenclamide (glyburide), while producing less prolonged insulin exposure and less risk of delayed hypoglycaemia. Further investigation is required to determine if nateglinide inhibition of postprandial glucose excursions will help to prevent diabetic complications or preserve pancreatic beta-cell function.
Product Description
Nateglinide, European Pharmacopoeia (EP) Reference Standard
USP
Nateglinide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Nateglinide, ≥98% (HPLC), solid