Skip to Content
MilliporeSigma
Nateglinide European Pharmacopoeia (EP) Reference Standard

Nateglinide

Nateglinide European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Fastic, N-[(trans-4-Isopropylcyclohexyl)carbonyl]-D-phenylalanine, Starlix, Starsis
Empirical Formula (Hill Notation):
C19H27NO3
CAS Number:
Molecular Weight:
317.42

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade