Skip to Content
MilliporeSigma
All Photos(1)

Documents

O7006-1AMP

Supelco

F.A.M.E. Mix RM-1

certified reference material, ampule of 100 mg (Neat)

Synonym(s):

AOCS Reference Mixtures, AOCS No. 1 (RM-1), Oil Reference Standard, AOCS No. 1, Oil Reference Standard, AOCS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 100 mg (Neat)
ampule of 100 mg fatty acid methyl ester mixture

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

functional group

carboxylic acid
ester

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Our quantitative animal and vegetable reference mixes conform to the requirements of the American Oil Chemists′ Society (AOCS) method Ce 1-62. The composition of each mix is similar to the fatty acid distribution of certain oils listed under each product. Please note that the AOCS mixtures sold under the Supelco brand are named RM-1, RM-2, etc.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
The composition of AOCS No. 1 (RM-1) is similar to the fatty acid distribution of the following oils: corn, cottonseed, kapok, poppyseed, rice, safflower, sesame, soybean, sunflower, and walnut oils. This mix may be used as a reference standard to aid in the identification and quantition of these oils.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Methyl arachidate 3 wt. %
    Methyl linoleate 50 wt. %
    Methyl linolenate 3 wt. %
    Methyl oleate 35 wt. %
    Methyl palmitate 6 wt. %
    Methyl stearate 3 wt. %

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service