Skip to Content
MilliporeSigma
All Photos(1)

Documents

O7756

Supelco

AOCS Low Erucic Rapeseed Oil

analytical standard, ampule of 100 mg

Synonym(s):

AOCS Reference Mixtures, F.A.M.E. Mix Rapeseed Oil, Rapeseed oil mixture, Oil Reference Standard, AOCS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

CofA

current certificate can be downloaded

packaging

ampule of 100 mg fatty acid methyl ester mixture
ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

shipped in

dry ice

storage temp.

-10 to -25°C

Looking for similar products? Visit Product Comparison Guide

General description

Our quantitative animal and vegetable reference mixes conform to the requirements of the American Oil Chemists′ Society (AOCS) method Ce 1-62. The composition of each mix is similar to the fatty acid distribution of certain oils listed under each product. Please note that the AOCS mixtures sold under the Supelco brand are named RM-1, RM-2, etc.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Methyl arachidate 3 wt. %
    Methyl behenate 3 wt. %
    Methyl cis-13-docosenoate 5 wt. %
    Methyl cis-11-eicosenoate 1 wt. %
    Methyl linoleate 12 wt. %
    Methyl linolenate 5 wt. %
    Methyl myristate 1 wt. %
    Methyl oleate 60 wt. %
    Methyl palmitate 4 wt. %
    Methyl stearate 3 wt. %
    Methyl tetracosanoate 3 wt. %
    See All (11)

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service