![F.A.M.E. Mix RM-1 certified reference material, ampule of 100 mg (Neat)](/deepweb/assets/sigmaaldrich/product/images/157/413/c943ae00-3544-4903-a77f-ea82e531c916/640/c943ae00-3544-4903-a77f-ea82e531c916.jpg)
F.A.M.E. Mix RM-1
![F.A.M.E. Mix RM-1 certified reference material, ampule of 100 mg (Neat)](/deepweb/assets/sigmaaldrich/product/images/157/413/c943ae00-3544-4903-a77f-ea82e531c916/640/c943ae00-3544-4903-a77f-ea82e531c916.jpg)
Synonym(s):
AOCS Reference Mixtures, Oil Reference Standard, AOCS
Products
Product Category
Brand
Product Line
Format
Quality Segment
Functional Group
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing