Skip to Content
MilliporeSigma
All Photos(1)

Documents

G005786

LC/MS/MS Analysis of Spice Cannabinoids in Plasma on Ascentis® Express F5 after Solid Phase Extraction (SPE) using HybridSPE®-Phospholipid

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

sample/matrix: rabbit plasma, unfiltered K2-EDTA spiked with Spice cannabinoids (5 ng/mL each)
SPE well plate: HybridSPE-Phospholipid, 96-well plate, 50 mg bed wt., 2 mL well vol (575656-U)
sample addition: to each well add 100 μL plasma, followed by a 300 μL of 1% formic acid in acetonitrile, agitate on orbital shaker for 2 minutes
elution: attach collection plate and apply vacuum at 10” Hg for 4 minutes
column: Ascentis Express F5, 5 cm x 2.1 mm I.D., 2.7 μm particles (53567-U)
mobile phase: [A] 10 mM ammonium formate in water, pH 6.8 (unadjusted); [B] acetonitrile; (50:50; A:B)
flow rate: 0.3 mL/min
pressure: 1296 psi (89 bar)
column temp.: 35 °C
detector: MS, ESI(+), MRM, m/z 344/155 (JWH-073 metabolite), 385/155 (JWH-200), 336/121 (JWH-250), 328/155 (JWH-073), and 342/155 (JWH-018)
injection: 2 μL

suitability

application for HPLC

application(s)

forensics and toxicology

Analysis Note

Synthetic cannabinoids (Spice) are a relatively new type of designer drug used as a pseudo-legal means to get a cannabis-type high. New synthetic cannabinoids are continually being introduced as suppliers tweak the molecular structures. The ability to rapidly and reliably identify the continually changing population of these compounds in the blood or urine suspected users is a significant analytical challenge facing forensic chemists. A four prong approach using column selectivity, high purity solvents, effective sample prep, and reference standards, was used to develop a method to rapidly isolate and identify Spice cannabinoids from plasma. The Ascentis Express F5 column gave the necessary resolution, and the highest grade LC-MS solvents and additives gave adduct-free response for maximum sensitivity. Sample prep employing the HybridSPE-Phospholipid was rapid and effective and the Cerilliant reference standards enabled confident identification.

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany
HybridSPE is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    JWH-073 N-(3-hydroxybutyl) metabolite solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    JWH-073 solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
    JWH-018 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service