Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

814449

TSKgel® Boronate-5PW (10 µm) HPLC Columns

L x I.D. 5 cm × 5 mm, m-aminophenyl boronate phase

Synonym(s):

TSKgel® Boronate-5PW HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

product name

TSKgel® Boronate-5PW HPLC Column, phase m-aminophenyl boronate, L × I.D. 5 cm × 5 mm, 10 μm particle size, glass

material

glass

product line

TSKgel®

analyte chemical class(es)

proteins

manufacturer/tradename

Tosoh 14449

parameter

≤20% organic solvent
≤3 M salt concentration
>0 °C temperature
1.2 mL/min flow rate
30 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

5 cm × 5 mm

matrix

hydroxylated methacrylic polymer

matrix active group

m-aminophenyl boronate phase

particle size

10 μm

operating pH

2-9

capacity

40 μmol/mL, resin adsorption capacity (sorbitol)

separation technique

affinity

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

High efficiency, resin-based TSKgel affinity columns (10 μm particles) separate or purify many enzymes and other proteins.

Coupling of m-aminophenyl boronate to TSKgel 5PW polymeric support results in a ligand capable of forming a tetrahedral boronate anion under alkaline pH conditions. This anionic structure can bind with 1,2 cis-diol groups such as those found in carbohydrates, carbohydrate containing compounds and catecholamines. Interaction between the boronate anion and the 1,2 cis-diol groups is enhanced in the presence of Mg2+ ions and is inhibited by amine-containing buffers. Adsorption onto the TSKgel Boronate-5PW matrix takes place in basic buffers such as HEPES and morpholine while desorption takes place in carbohydrate or amine-containing mobile phase like sorbitol or Tris.

Applications include: nucleic acids, catecholamines, nucleosides and nucleotides.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service