Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

823483

TSKgel® Protein A-5PW (20 µm) HPLC Columns

L x I.D. 3.5 cm × 4.6 mm, Protein A phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709

Product Name

TSKgel® Protein A-5PW HPLC Column, 20 μm particle size, L × I.D. 3.5 cm × 4.6 mm

material

PEEK

product line

TSKgel®

manufacturer/tradename

TSKgel®

parameter

20 bar pressure (290 psi)
4-30 °C temperature

technique(s)

HPLC: suitable

L × I.D.

3.5 cm × 4.6 mm

matrix

polymeric particle platform

matrix active group

Protein A phase

particle size

20 μm

separation technique

affinity

storage temp.

2-8°C

General description

TSKgel Protein A-5PW is a 20 μm, 4.6 mm ID × 3.5 cm column for high performance affinity chromatography. Made of PEEK hardware, this column has been designed for the rapid separation and robust quantification of a
variety of antibodies. Monoclonal antibodies from harvested cell culture media can be captured and accurately quantitated in less than 2 minutes per injection.

The TSKgel Protein A-5PW column can be used for more than 2,000 injections without regeneration or cleaning. Packed with hydroxylated methacrylic polymer beads, this column is designed with a high degree
of crosslinking, which allows a high flow rate for chromatography while still maintaining chromatographic efficiency, peak width and resolution. The recombinant protein A ligand is a code-modified hexamer of the C domain.
An enhanced rProtein A ligand is bound to the TSKgel 5PW base bead via multipoint attachment resulting in excellent base stability in 0.1 mol/L NaOH.

Footnote

Highlights

  • Wide dynamic range with high sample concentration load for mAb quantitation
  • Fast analysis: 1-2 min/analysis
  • High sensitivity for mAb determination
  • Long lifetime: .2,000 injections per column with no sign of deterioration

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service