Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

666742

Sigma-Aldrich

3-Formyl-4-isopropoxyphenylboronic acid

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)CHOC6H5(CHO)B(OH)2
CAS Number:
Molecular Weight:
208.02
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.06

Quality Level

Assay

≥95%

mp

98-102 °C

functional group

aldehyde

storage temp.

2-8°C

SMILES string

CC(C)Oc1ccc(cc1C=O)B(O)O

InChI

1S/C10H13BO4/c1-7(2)15-10-4-3-9(11(13)14)5-8(10)6-12/h3-7,13-14H,1-2H3

InChI key

MFWMHBLPARKSLU-UHFFFAOYSA-N

Application

  • Used as a lubricating additive in grease for reducing fretting wear

Other Notes

May contain varying amounts of anhydride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service