Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

808806

TSKgel® DEAE-5PW (20 µm) HPLC Columns

L x I.D. 1 cm × 8 mm, DEAE (diethylaminoethyl) phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

product name

TSKgel® DEAE-5PW Guards and Guardgel Kits, phase DEAE (diethylaminoethyl), L × I.D. 1 cm × 8 mm, 20 μm particle size, glass

material

glass

product line

TSKgel®

feature

counter ion Cl-

manufacturer/tradename

Tosoh 08806

parameter

≤1 M salt concentration
≤20% organic solvent
10-45 °C temperature

technique(s)

LPLC: suitable

L × I.D.

1 cm × 8 mm

matrix

hydroxylated methacrylic polymer

matrix active group

DEAE (diethylaminoethyl) phase

particle size

20 μm

operating pH

2-12

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

A TSKgel® Guardgel Kit contains 5 mL of the packing material, specified in the product description, in an appropriate slurry solvent. In the case of a glass Guardgel Kit, the product also includes a column holder, a glass column, 10 mm L x 13 mm OD x 8 mm ID, 2 external endfittings, 2 caps with M6 threads, and a packing adaptor. Simple instructions to pack the guard column are included in the shipment.
TSKgel Guard cartridges require a holder, which is sold separately. For 2 mm cartridges, the holder is Part No. 819308. For the 3.2 mm cartridges used to protect 4.6 mm columns, the holder is Part No. 819018.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service