Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18800

Sigma-Aldrich

Octanedioic acid

for synthesis

Synonym(s):

Octanedioic acid, Suberic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOC(CH2)6COOH
CAS Number:
Molecular Weight:
174.19
MDL number:
UNSPSC Code:
12352106
EC Index Number:
208-010-9
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

<1 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

autoignition temp.

430 °C

potency

>2000 mg/kg LD50, oral (Rat)

pH

3.2 (20 °C, 1.6 g/L in H2O)

bp

229-231 °C/20 hPa

mp

139-142 °C

transition temp

flash point 210 °C

bulk density

700 kg/m3

storage temp.

2-30°C

SMILES string

[O-]C(=O)CCCCCCC(=O)[O-].[H+].[H+]

InChI

1S/C8H14O4/c9-7(10)5-3-1-2-4-6-8(11)12/h1-6H2,(H,9,10)(H,11,12)

InChI key

TYFQFVWCELRYAO-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 139 °C
Melting range (upper value): ≤ 142 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

410.0 °F - closed cup

Flash Point(C)

210 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service