Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

400599

Sigma-Aldrich

Rubidium chromate

Synonym(s):

Dioxido(dioxo)chromium rubidium(1+)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Rb2CrO4
CAS Number:
Molecular Weight:
286.93
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

form

powder

Quality Level

concentration

14.5-21.7% Cr (by Na2S2O3, titration)

density

3.518 g/mL at 25 °C (lit.)

SMILES string

[Rb+].[Rb+].[O-][Cr]([O-])(=O)=O

InChI

1S/Cr.4O.2Rb/q;;;2*-1;2*+1

InChI key

WXLJFWCCZKCVSQ-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Ox. Sol. 2 - Skin Sens. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service