Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

79322

Supelco

Extractables and Leachables PE/PP mix for LC

certified reference material, TraceCERT®, 13 compounds, in acetonitrile, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.21
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

in acetonitrile

application(s)

pharmaceutical

format

multi-component solution

storage temp.

−20°C

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from an NMI, e.g. NIST, NRC or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Check out our complete range of extractables and leachables certified reference mixes

Preparation Note

For the preparation of this CRM only materials of highest purity were used.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Tris(3,5-di-tert-butyl-4-hydroxybenzyl) isocyanurate
    2,4-Di-tert-butylphenol
    Octadecyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate
    Didodecyl 3,3′-thiodipropionate
    Bis(2-ethylhexyl) phthalate
    Palmitic acid
    Stearic acid
    Oleic acid
    Pentaerythritol tetrakis(3,5-di-tert-butyl-4-hydroxyhydrocinnamate)
    Tris(2,4-di-tert-butylphenyl)phosphate
    cis-13-Docosenoamide
    Ethyl 4-ethoxybenzoate
    Bisphenol A
    See All (13)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - (Acetonitrile)

Flash Point(C)

2.0 °C - (Acetonitrile)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service