Skip to Content
MilliporeSigma

01829

Extractables and Leachables Screening Standard for GC

certified reference material, TraceCERT®, 14 components in tert-Butyl methyl ether, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

certified reference material, TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

50 mg/L each component (nominal concentration)

application(s)

chemicals and industrial polymers
food and beverages
pharmaceutical

format

multi-component solution

storage temp.

−20°C

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from an NMI, e.g. NIST, NRC or NMIJ.


Certified content incl. uncertainty and expiry date are given on the certificate.


Download your certificate at: http://www.sigma-aldrich.com.

Check out our complete range of extractables and leachables certified reference mixes




Discover our new Extractables and Leachables Silicone Mix with the most commonly found Silicone extractables

Application

Extractables Study with GC/MS - Extractables Studies of Single-Use Equipment article from >AnalytixReporter issue 8

Preparation Note

For the preparation of this CRM only materials of highest purity were used.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany


Analyte

Description

  • Octadecyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate

  • ε-Caprolactam

  • 2-Mercaptobenzothiazole

  • Bisphenol A

  • 3,5-Di-tert-butyl-4-hydroxytoluene

  • 1,3-Di-tert-butylbenzene

  • Oleamide

  • Bis(2-ethylhexyl) phthalate

  • Stearic acid

  • cis-13-Docosenoamide

  • Tris(2,4-di-tert-butylphenyl)phosphate

  • 2,4-Di-tert-butylphenol

  • 2,6-Di-tert-butylphenol

  • Palmitic acid

See All (14)

pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

-18.4 °F - closed cup

flash_point_c

-28 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library