Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

59271

SSI High Pressure Precolumn Filter

10-32 Waters thread

Synonym(s):

COLUMN PREFILTER,WATERS FITTING, 0.5UM, EA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

packaging

pkg of 1 ea

fitting

10-32 Waters thread

Looking for similar products? Visit Product Comparison Guide

General description

In-line installation. The 316 stainless steel filter disc (0.5 μm pores) is easily replaced without removing the column end fitting. For 1/16 in. tubing. Maximum operating pressure: 15,000 psi (1,054 kg/cm2).

Components

nuts and ferules for 1/16 in. tubing included

Legal Information

SSI is a trademark of Scientific Systems, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service