Key Documents
59269
SSI High Pressure Precolumn Filter
10-32 Valco compatible thread
Synonym(s):
COLUMN PREFILTER, CPI FITTING,0.5UM, EA
About This Item
UNSPSC Code:
41115700
packaging
pkg of 1 ea
fitting
10-32 Valco compatible thread
General description
In-line installation. The 316 stainless steel filter disc (0.5 μm pores) is easily replaced without removing the column end fitting. For 1/16 in. tubing. Maximum operating pressure: 15,000 psi (1,054 kg/cm2).
Legal Information
SSI is a trademark of Scientific Systems, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.