Key Documents
58827
Rheodyne® Model 7010 Injector
About This Item
UNSPSC Code:
41115854
feature
number of ports 6
positions 2
General description
The original Rheodyne HPLC injector, Model 7010 introduces 5 μL - 5 mL samples onto a column with excellent reproducibility (intended for complete loop filling only). Six clustered ports allow compact interconnections, minimizing tubing volume. The flat-face seal is easy to adjust and service. Operates at pressures to 7000 psi (490 kg/cm2). Supplied with a 20 μL loop; order fill port separately.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Rheodyne is a registered trademark of IDEX Health & Science, LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)