Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z226688

PEEK Tubing

L × O.D. × I.D. 10 ft × 1/16 in. × 0.007 in., yellow, max. pressure 8000

Synonym(s):

PEEK Tubing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32119290
NACRES:
SB.52

parameter

8000 max. pressure

L × O.D. × I.D.

10 ft × 1/16 in. × 0.007 in.

color

yellow

Looking for similar products? Visit Product Comparison Guide

General description

Polyetheretherketone (PEEK) tubing has become a popular replacement for stainless steel tubing in various places in the HPLC system. It is especially useful when contact between the sample and metal components must be avoided, such as when working with metal complexing agents or certain biochemical compounds. PEEK tubing also offers other advantages: it is flexible, easy to cut, has excellent mechanical stability and chemical compatibility, and can be used to 100 °C. Tetrahydrofuran (THF), dimethyl sulfoxide (DMSO), methylene chloride, and concentrated nitric and sulfuric acids should not be used with PEEK tubing. Our 1/16 in. PEEK tubing is color coded for easy identification of the internal diameter. 10 ft./3 m length.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service