Key Documents
57621
Rheodyne® Injector
Rheodyne 7725i
About This Item
UNSPSC Code:
41115854
description
internal position sensing switch
feature
manual
number of ports 6
positions 2
manufacturer/tradename
Rheodyne 7725i
parameter
7000 psi max. pressure
General description
Rheodyne® Injector model is a stainless steel version and comprises of a six-port sample injection valves in which the sample is loaded through a built-in needle port in the front of the valve.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Rheodyne is a registered trademark of IDEX Health & Science, LLC
recommended
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.