Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

58697-U

PTFE Tubing

L × O.D. × I.D. 50 ft × 1/16 in. × 0.023 in.

Synonym(s):

PTFE Tubing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32119290
NACRES:
SB.52

parameter

500 psi max. pressure

L × O.D. × I.D.

50 ft × 1/16 in. × 0.023 in.

Looking for similar products? Visit Product Comparison Guide

General description

Economical, flexible PTFE tubing is ideal for use at pressures up to 500 psi (35 kg/cm2). Use in automatic analyzer, postcolumn reaction, preparative scale systems, in-stream sampling devices, and when monitoring physiologically important compounds.

The maximum recommended operating temperature for PTFE is 200 °C, but short term exposure to higher temperatures seldom causes damage.

Application


  • Naked-eye sensitive detection of nanoPET by pH-responsive colorimetric method based on dual-enzyme catalysis: This study employs Bromocresol Purple for the sensitive detection of pollutants, utilizing its pH-responsive colorimetric properties in a novel environmental monitoring approach, which is crucial for detecting nano-pollutants in environmental samples (Zhan et al., 2024).

  • Noninvasive Colorimetric Detection of Acetic Acid in Human Breath Based on an Alginate/Ni-Al-LDH/Dye Composite Film: Integrates Bromocresol Purple in a bio-composite for non-invasive detection of acetic acid, showcasing its potential in medical diagnostics and monitoring of metabolic disorders (Zakaria et al., 2023).

  • Fish Freshness Indicator for Sensing Fish Quality during Storage: Uses Bromocresol Purple as a colorimetric indicator to assess fish freshness, offering a valuable tool for ensuring food safety and quality in the seafood industry (Kim et al., 2023).

  • An Adapted Protocol for Quantitative Rhizosphere Acidification Assay: Highlights the application of Bromocresol Purple in ecological research, particularly in studying soil health and plant interactions, which is vital for agricultural sciences (Omer et al., 2023).

  • Halochromic Textiles for Real-Time Sensing of Hazardous Chemicals and Personal Protection: Demonstrates the use of Bromocresol Purple in developing halochromic textiles, which change color in response to chemical exposures, crucial for safety in hazardous environments (Leite et al., 2023).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service