Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

57276

Supelco

1/8" PTFE tubes, Color Coded

for use with Supelco Visiprep Large Volume Sampler, pk of 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

feature

color-coded

packaging

pk of 4

technique(s)

solid phase extraction (SPE): suitable

tubing O.D.

1/8 in.

application(s)

food and beverages

compatibility

for use with Supelco Visiprep Large Volume Sampler


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service