Skip to Content
MilliporeSigma
All Photos(1)

Documents

57209

Supelco

Supelclean LC-Florisil® SPE Bulk Packing

pkg of 100 g

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41115712

product line

Supelclean

Quality Level

packaging

pkg of 100 g

technique(s)

solid phase extraction (SPE): suitable

matrix

magnesium silicate

matrix active group

Florisil phase

diameter

100-120 mesh

application(s)

food and beverages

separation technique

normal phase

General description

LC-Florisil Supelclean SPE tubes may be used to extract polar analytes from nonpolar samples.

Legal Information

Florisil is a registered trademark of U.S. Silica Company
Supelclean is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service