Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47995-U

Supelco

Di(2-ethylhexyl)adipate solution

certified reference material, 2000 μg/mL in methanol

Synonym(s):

Bis(2-ethylhexyl) adipate, DEHA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Agency

EPA 506

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

2000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

O(CC(CCCC)CC)C(=O)CCCCC(=O)OCC(CCCC)CC

InChI

1S/C22H42O4/c1-5-9-13-19(7-3)17-25-21(23)15-11-12-16-22(24)26-18-20(8-4)14-10-6-2/h19-20H,5-18H2,1-4H3

InChI key

SAOKZLXYCUGLFA-UHFFFAOYSA-N

General description

Di(2-ethylhexyl)adipate is an environmental pollutant belonging to the class of phthalate esters. It is extensively used as an additive in the manufacturing of polyvinyl chloride (PVC) plastics which aid in adding flexibility.

Application

Di(2-ethylhexyl)adipate may be used as a reference standard for the quantification of the analyte in food packaging films and water samples using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of phthalate esters in water samples by solid-phase microextraction and gas chromatography with mass spectrometric detection
Penalver.A, et al.
Journal of Chromatography A, 872(1-2), 191-201 (2000)
Extraction and gas chromatographic evaluation of plasticizers content in food packaging films
Bonini M, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 90(1), 31-36 (2008)
Mohd Basyaruddin Abdul Rahman et al.
Applied biochemistry and biotechnology, 158(3), 722-735 (2009-01-10)
In this study, an artificial neural network (ANN) trained by backpropagation algorithm, Levenberg-Marquadart, was applied to predict the yield of enzymatic synthesis of dioctyl adipate. Immobilized Candida antarctica lipase B was used as a biocatalyst for the reaction. Temperature, time
Kyoko Nabae et al.
Reproductive toxicology (Elmsford, N.Y.), 22(3), 411-417 (2006-09-06)
F344 male rats were given five consecutive weekly subcutaneous injections of folic acid for induction of chronic renal dysfunction and then di(2-ethylhexyl)phthalate (DEHP) or di(2-ethylhexyl)adipate (DEHA) in the diet at a concentration of 0, 6000 or 25,000 ppm for 4
Julie Borch et al.
Reproductive toxicology (Elmsford, N.Y.), 19(4), 517-525 (2005-03-08)
This study aimed to characterize the effects of di(2-ethylhexyl) phthalate (DEHP) on the fetal rat testes and relate them to the effects seen in adults. Histopathological effects in fetal testes were examined with immunohistochemistry for anti-Mullerian hormone (AMH), 3beta-hydroxysteroid dehydrogenase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service