Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

450588

Sigma-Aldrich

Diisobutyl adipate

99%

Synonym(s):

Hexanedioic acid 1,6-bis(2-methylpropyl) ester, NSC 6343

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CH2CO2CH2CH(CH3)2]2
CAS Number:
Molecular Weight:
258.35
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

refractive index

n20/D 1.432 (lit.)

bp

293 °C (lit.)

density

0.954 g/mL at 25 °C (lit.)

SMILES string

CC(C)COC(=O)CCCCC(=O)OCC(C)C

InChI

1S/C14H26O4/c1-11(2)9-17-13(15)7-5-6-8-14(16)18-10-12(3)4/h11-12H,5-10H2,1-4H3

InChI key

RDOFJDLLWVCMRU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diisobutyl adipate is a bis(2-methylpropyl) ester of adipic acid that is used as a plasticizer and a fatty acid in synthetic oils.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of plasticisers in food by gas chromatography-mass spectrometry with ion-trap mass detection.
Lau O and Wong S
Journal of Chromatography A, 737(2), 338-342 (1996)
Synthetic ester-based oils and their application in power industry.
International Conference on Renewable Energies and Power Quality (ICREPQ?09) (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service