Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47212

Supelco

ASTM® D4815 Valve Timing Mix

(each component at the nominal wt% indicated), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

analytical standard

Agency

ASTM® D4815

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

(each component at the nominal wt% indicated)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

2-30°C

General description

ASTM® D4815 Valve Timing Mix may be used for the determination of oxygenates (ethers and alcohols) in gasoline by GC. All components used in preparing the standards have been analyzed for purity, water content, and the presence of other method components to 0.01%. Blends have been prepared using precise gravimetric techniques exceeding the requirements of ASTM Method D4815. Corrections are made for common impurities. All calibration blends are provided with a chromatogram and data verifying the purity and identity of the raw materials.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Each component at the nominal weight percent listed. Use for determining the proper backflushing valve timing.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

    tert-Butyl ethyl ether 10 % (w/w)
    tert-Butyl methyl ether 10 % (w/w)
    Diisopropyl ether 10 % (w/w)
    Hexane 60 % (w/w)
    Methylcyclopentane 10 % (w/w)

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-9.4 °F - closed cup

Flash Point(C)

-23 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service