Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47211

Supelco

ASTM® D4815 Quantitative Calibration Kit

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

analytical standard

Agency

ASTM® D4815

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

2-30°C

General description

ASTM® D4815 Quantitative Calibration Kit may be used for the determination of oxygenates (ethers and alcohols) in gasoline by GC. All components used in preparing the standards have been analyzed for purity, water content, and the presence of other method components to 0.01%. Blends have been prepared using precise gravimetric techniques exceeding the requirements of ASTM Method D4815. Corrections are made for common impurities. All calibration blends are provided with a chromatogram and data verifying the purity and identity of the raw materials.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Each of the following products.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Kit Components Also Available Separately

Product No.
Description
SDS

  • 47205D4815 Quantitative Calibration Mix 1 (47205)SDS

  • D4815 Quantitative Calibration Mix 2 (47206)

  • 47208D4815 Quantitative Calibration Mix 4 (47208)SDS

  • 47209D4815 Quantitative Calibration Mix 5 (47209)SDS

  • 47212D4815 Valve Timing Mix (47212)SDS

  • 47213D4815 Qualitative ID Mix (47213)SDS

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Carc. 1A - Eye Dam. 1 - Flam. Liq. 2 - Muta. 1B - Repr. 1B - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT RE 2 - STOT SE 2 - STOT SE 3

Target Organs

Blood, Central nervous system, Eyes,Central nervous system, Nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

Flash Point(F)

-27.4 °F

Flash Point(C)

-33 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Complete offer for petrochemical testing including environmental and industrial hygiene monitoring solutions.

Complete offer for petrochemical testing including environmental and industrial hygiene monitoring solutions.

Complete offer for petrochemical testing including environmental and industrial hygiene monitoring solutions.

Complete offer for petrochemical testing including environmental and industrial hygiene monitoring solutions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service