Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

87612

Supelco

Amaranth

analytical standard

Synonym(s):

Acid Red 27, Azorubin S, FD & C Red Dye No. 2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H11N2Na3O10S3
CAS Number:
Molecular Weight:
604.47
Colour Index Number:
16185
Beilstein:
4122311
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
E Number:
E123
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[Na+].[Na+].[Na+].Oc1c(N=Nc2ccc(c3ccccc23)S([O-])(=O)=O)c4ccc(cc4cc1S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C20H14N2O10S3.3Na/c23-20-18(35(30,31)32)10-11-9-12(33(24,25)26)5-6-13(11)19(20)22-21-16-7-8-17(34(27,28)29)15-4-2-1-3-14(15)16;;;/h1-10,23H,(H,24,25,26)(H,27,28,29)(H,30,31,32);;;/q;3*+1/p-3

InChI key

WLDHEUZGFKACJH-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

417.2 °F - closed cup

Flash Point(C)

214 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shekhar B Jadhav et al.
Environmental science and pollution research international, 20(5), 2854-2866 (2012-09-22)
Bacterium Pseudomonas aeruginosa BCH was able to degrade naphthylaminesulfonic azo dye Amaranth in plain distilled water within 6 h at 50 mg l(-1) dye concentration. Studies were carried out to find the optimum physical conditions and which came out to
Guowen Zhang et al.
Food chemistry, 136(2), 442-449 (2012-11-06)
The mechanism of interaction between food dye amaranth and human serum albumin (HSA) in physiological buffer (pH 7.4) was investigated by fluorescence, UV-vis absorption, circular dichroism (CD), and Fourier transform infrared (FT-IR) spectroscopy. Results obtained from analysis of fluorescence spectra
Vinod K Gupta et al.
Materials science & engineering. C, Materials for biological applications, 32(1), 12-17 (2012-11-28)
The photo-catalytic degradation of an azo dye - Amaranth (AM) - has been investigated in TiO(2)/UV aqueous suspensions. The results obtained from the experiments during H(2)O(2)/TiO(2) addition show that the highest decolorization rate is provided by the combination of (UV+TiO(2)+H(2)O(2)).
S Nadupalli et al.
The journal of physical chemistry. A, 115(27), 7948-7954 (2011-06-09)
The reaction mechanism of the oxidation of Amaranth dye (2-hydroxy-1-(4-sulfonato-1-naphthylazo) naphthalene-3,6-disulfonate) with hypochlorite under varied pH conditions was elucidated by a kinetic approach. Under excess concentration of oxidant, the reaction followed pseudo-first-order kinetics with respect to Amaranth, and the oxidation
Yuanyuan Zhang et al.
Analytica chimica acta, 764, 53-58 (2013-02-05)
Different-shaped aluminas were readily prepared via hydrothermal reaction. It was found that the morphology and the electrochemical sensing properties of alumina were heavily dependent on the reaction time. When extending the reaction time from 6 h to 24 h, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service