Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40900-U

Supelco

Transformer oil (PCB free)

Synonym(s):

Non-PCB Insulating Oil

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

analytical standard

CofA

current certificate can be downloaded

packaging

pkg of 250 mL

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

matrix material

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

Transformer oil (PCB free) is a neat reference standard for environmental analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Transformer oil (PCB free) has been used as an analytical standard for the quantification of polychlorinated biphenyls in transformer oil samples using gas chromatography coupled to mass spectrometry (GC-MS).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

294.8 °F - closed cup

Flash Point(C)

146 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Determination of polychlorinated biphenyls in transformer oil using various adsorbents for solid phase extraction
Na C-Y, et al.
Chemosphere, 73(1), S7-S12 (2008)
Screening method for rapid determination of polychlorinated biphenyls in transformer oil by liquid-liquid extraction and gas chromatography-mass spectrometry
Toledo C, et al.
Journal of the Brazilian Chemical Society, 18(5), 937-942 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service