Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

46956

Supelco

Transformer oil (PCB free)

analytical standard, PCB free

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502

grade

analytical standard

CofA

current certificate can be downloaded

packaging

pkg of 10 × 5 mL

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

matrix material

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

Sigma-Aldrich offers transformer oil (PCB free) for a variety of applications that include, but are not limited to, environmental, food and beverage, forensics, petrochemical and pharmaceutical analyses. Purity of the chemical is about 99% unless otherwise stated.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

294.8 °F - closed cup

Flash Point(C)

146 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service