CRMUPPGO
Pour Point Standard
IP15, ASTM D97, ISO 3016, BS 2000, Pt 15 (-34 °C)
Synonym(s):
IP15, ASTM D97, Pt 15(-34 °C)Pour Point Standard
About This Item
Recommended Products
Agency
according to ASTM® D97
according to BS 2000
according to ISO 3016
manufacturer/tradename
Paragon Scientific Ltd
bp
141-462 °C
application(s)
petroleum
format
mixture
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
134.6 °F
Flash Point(C)
57 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service