Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

28271-U

Supelco

Tenax® TA Thermal Desorption Tube

greener alternative

stainless steel, O.D. x I.D. x L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea

Synonym(s):

Tenax TA Thermal Desorption Tube (stainless steel)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

Product Name

Tenax® TA, O.D. × I.D. × L 6 mm × 4 mm × 7 in., stainless steel TD tube, preconditioned, 60-80 mesh, pkg of 1 ea

material

stainless steel TD tube

Quality Level

Agency

EPA TO-1,IP-1B

description

Sealed with TDS³ Storage Container

feature

preconditioned

packaging

pkg of 1 ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

active air sampling: suitable

O.D. × I.D. × L

6 mm × 4 mm × 7 in.

matrix

Tenax TA porous polymer

particle size

60-80 mesh

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with Gerstel® TDS A & TDS 2

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

Tenax® TA is a prepacked, stainless steel sampling tube that is fully compatible with the GERSTEL instrument. It employs a stainless steel screen at both inlet and outlet to keep the adsorbent beds secure during use.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. Thermal desorption tubes are reusable at least 50 to 100 times, and reduces the usage of accessories like transfer vials, caps, and pipette tips, thus aligns with “Waste Prevention”. Also, these tubes eliminate the usage of toxic solvents like CS2 to extract the VOCs of interest, and thus aligns with “Safer Solvents and Auxiliaries” principle. Click here for more information.

Application


  • Chemical Composition of Atmospheric Air in Nemoral Scots Pine Forests and Submountainous Beech Forests: The Potential Region for the Introduction of Forest Therapy.: This study analyzes the air quality in forest areas, potentially highlighting the use of Tenax® TA in environmental monitoring for forest therapy research (Dudek et al., 2022 doi:10.3390/ijerph192315838).

  • Multivariate optimization of dual-sorbent dynamic headspace extraction of volatiles in wine analysis.: Focuses on optimizing headspace extraction techniques in the analysis of wine volatiles, illustrating the practical applications of Tenax® TA in food chemistry and analytical methods (Vyviurska et al., 2021 doi:10.1016/j.foodchem.2021.130449).

  • Impact of the adsorbent material on volatile metabolites during in vitro and in vivo bio-sampling.: Investigates how adsorbent materials like Tenax® TA affect the collection and analysis of volatile organic compounds, which is crucial for accurate metabolic profiling (Franchina et al., 2021 doi:10.1016/j.talanta.2020.121569).

  • Challenges of fast sampling of volatiles for thermal desorption gas chromatography - mass spectrometry.: Discusses the challenges and technical considerations in the rapid sampling of volatiles, relevant for chemists using Tenax® TA in high-throughput analytical settings (Marcillo et al., 2020 doi:10.1016/j.chroma.2019.460822).

  • A green approach for the extraction of diamondoids from petroleum source rock.: Describes an environmentally friendly method using Tenax® TA for extracting complex hydrocarbons, useful in both academic research and industrial applications (Akinlua et al., 2019 doi:10.1016/j.aca.2019.08.071).

Legal Information

GERSTEL is a registered trademark of Gerstel GmbH & Co. KG
Tenax is a registered trademark of Buchem B.V.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service