Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

28281-U

Supelco

Tenax® TA Thermal Desorption Tube

greener alternative

glass, O.D. x I.D. x L 6 mm × 4 mm × 7 in., fritted, preconditioned, pkg of 1 ea

Synonym(s):

Tenax TA Thermal Desorption Tube (glass)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

Product Name

Tenax® TA, glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, 60-80 mesh, pkg of 1 ea

material

glass TD tube

Quality Level

Agency

EPA TO-1,IP-1B

description

Sealed with TDS³ Storage Container

feature

fritted
preconditioned

packaging

pkg of 1 ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

active air sampling: suitable

O.D. × I.D. × L

6 mm × 4 mm × 7 in.

matrix

Tenax TA porous polymer

particle size

60-80 mesh

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with Gerstel® TDS A & TDS 2

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

Each Tenax® TA sampling tube is individually numbered, thermally conditioned, and batch tested for back-pressure and background. These prepacked standard sampling tubes are sealed in exclusive storage containers. They incorporate a glass frit at the inlet, which increases their performance.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. Thermal desorption tubes are reusable at least 50 to 100 times, and reduces the usage of accessories like transfer vials, caps, and pipette tips, thus aligns with “Waste Prevention”. Also, these tubes eliminate the usage of toxic solvents like CS2 to extract the VOCs of interest, and thus aligns with “Safer Solvents and Auxiliaries” principle. Click here for more information.

Application


  • Use of PLE-ATD-GC/MSMS for the Quantification of Airborne Pesticides in Active and Passive Samples and in Dust.: This study leverages Pressurized Liquid Extraction (PLE) and Automatic Thermal Desorption (ATD) coupled with Gas Chromatography-Tandem Mass Spectrometry (GC/MSMS) for the quantification of airborne pesticides. Tenax TA was utilized to collect airborne particulates and VOCs from both active and passive air samples, illustrating its efficacy in capturing and analyzing environmental contaminants (Rodrigues et al., 2024).

  • Chemical Composition of Atmospheric Air in Nemoral Scots Pine Forests and Submountainous Beech Forests: The Potential Region for the Introduction of Forest Therapy.: This research analyzes the chemical composition of atmospheric air in specific forest environments, employing Tenax TA adsorbents to trap and analyze volatile organic compounds. The findings contribute to understanding the therapeutic potential of forest environments based on their VOC profiles (Dudek et al., 2022).

  • Volatiles of the Apicomplexan Alga Chromera velia and Associated Bacteria.: This study focuses on identifying volatile compounds from the alga Chromera velia and its associated bacteria, utilizing Tenax TA to collect and analyze the VOCs emitted. The research offers insights into the ecological interactions and chemical communication between the alga and bacteria (Koteska et al., 2023).

  • Real-world emission characteristics of semivolatile/intermediate-volatility organic compounds originating from nonroad construction machinery in the working process.: This study investigates the emission characteristics of organic compounds from construction machinery, using Tenax TA tubes to collect emissions during real-world operations. The analysis aids in assessing the environmental impact and occupational hazards associated with these emissions (Shen et al., 2023).

  • Identification of Volatiles of the Dinoflagellate Prorocentrum cordatum.: Focused on the dinoflagellate Prorocentrum cordatum, this research identifies various volatile compounds using Tenax TA, exploring the species′ potential impacts on marine ecosystems and its interactions within the marine microbial community (Koteska et al., 2022).

Legal Information

GERSTEL is a registered trademark of Gerstel GmbH & Co. KG
Tenax is a registered trademark of Buchem B.V.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service