Skip to Content
MilliporeSigma
All Photos(1)

Documents

21009-U

Supelco

Tenax® Porous Polymer Adsorbent

matrix Tenax TA, 80-100 mesh, bottle of 10g

Synonym(s):

polymeric adsorbent, XAD7, 20-60 mesh

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
NACRES:
SB.54

product name

Tenax® Porous Polymer Adsorbent, matrix Tenax TA, 80-100 mesh, bottle of 10 g

form

solid

Quality Level

packaging

bottle of 10 g

parameter

350 °C temp. limit

surface area

~35 m2/g

matrix

Tenax TA

particle size

80-100 mesh

density

~0.29 g/mL (free fall density)

Looking for similar products? Visit Product Comparison Guide

General description

Porous polymer sorbents are inert, hydrophobic with large surface area. These sorbents retain volatile compounds, water and solvent vapours. This is advantageous if sample is from atmosphere with high content of water and solvent vapours. They are most suitable for trapping toxic agents with high molecular weight and non-volatile substances like pesticides. They are also suitable for sampling many compounds sorbed by charcoal and silica gel. Tenax® TA is basically synthetically produced polymer based on the 2,6-diphenylene oxide is more efficient in the recovery of higher molecular weight compounds. It is suitable for both polar and nonpolar analytes.
Tenax porous polymers, based on 2,6-diphenyl-p-phenylene oxide, is widely used as an adsorbent in both air collection and purge trap applications. Its unique structure provides alternate and desirable adsorption/desorption characteristics compared to other porous polymers.

For more information about any of our adsorbents, please visit sigma-aldrich.com/adsorbents

Application

Tenax® TA is suitable as an adsorbent used for air sampling, it was examined for its potential to form artifacts with Ozone at environmental concentrations, which was then quantitatively analysed using GC-MS analysis. It is also used in adsorbent columns suitable for recovering volatile compounds analysed by GC.

Legal Information

Tenax is a registered trademark of Buchem B.V.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Production and recovery of aroma compounds produced by solid-state fermentation using different adsorbents.
Medeiros, Adriane BP, et al.
Food Technology and Biotechnology, 44, 47-51 (2006)
Niamh Nic Daeid
Fire Investigation , 143-144 (2004)
V.G. Berezkin, Yu.S. Drugov
Gas Chromatography in Air Pollution Analysis, 50-50 (1991)
Vânia F Pais et al.
Sensors (Basel, Switzerland), 12(2), 1422-1436 (2012-03-23)
An electronic nose based on coated piezoelectric quartz crystals was used to distinguish cheese made from ewes' milk, and to distinguish cheese varieties. Two sensors coated with Nafion and Carbowax could certify half the ewes' cheese samples, exclude 32 cheeses
Jae Hwan Lee et al.
Journal of the Air & Waste Management Association (1995), 56(11), 1503-1517 (2006-11-23)
Four popular thermally desorbable adsorbents used for air sampling (Tenax TA, Tenax GR, Carbopack B, and Carbopack X) are examined for the potential to form artifacts with ozone (O3) at environmental concentrations. The performance of these adsorbents for the ketone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service