Key Documents
2633601
Supelco
Inlet Liner, Splitless Type, Straight Design (unpacked)
pkg of 1 ea
About This Item
UNSPSC Code:
41115719
packaging
pkg of 1 ea
manufacturer/tradename
Shimadzu 221-41444-01 (OEM)
L × O.D. × I.D.
95 mm × 5.0 mm × 3.4 mm
compatibility
technique used for General purpose
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Shimadzu (17A) [with SPL-17 Injector]
Choose from one of the most recent versions:
Certificates of Analysis (COA)