Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

2637505

Supelco

Inlet Liner, Direct (SPME) Type, Straight Design (unpacked)

pkg of 5 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

packaging

pkg of 5 ea

L × O.D. × I.D.

78.5 mm × 6.5 mm × 0.75 mm

Looking for similar products? Visit Product Comparison Guide

General description

Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Agilent (5890, 6890, and 7890)
  • Fits Finnigan (9001GCQ)
  • Fits Varian (CP-1177 Injector)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fire Debris Analysis.
Stauffer E
Science, 249-252 (2007)

Articles

The work describes the development of an HS-SPME-GC-MS method to determine 2-chloroethanol as a marker of fumigant ethylene oxide in different kinds of sesame seed samples.

The work describes the development of an HS-SPME-GC-MS method to determine 2-chloroethanol as a marker of fumigant ethylene oxide in different kinds of sesame seed samples.

The work describes the development of an HS-SPME-GC-MS method to determine 2-chloroethanol as a marker of fumigant ethylene oxide in different kinds of sesame seed samples.

The work describes the development of an HS-SPME-GC-MS method to determine 2-chloroethanol as a marker of fumigant ethylene oxide in different kinds of sesame seed samples.

See All

Protocols

In this article, SPME-GC was successfully used to identify many volatile compounds present in the headspace over several spirits.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service