Key Documents
2631105
Supelco
Inlet Liner, Splitless Type, Straight Design (unpacked)
pkg of 5 ea
About This Item
UNSPSC Code:
12000000
description
Splitless for AutoSystem
packaging
pkg of 5 ea
manufacturer/tradename
PerkinElmer N610-1372 (OEM)
PerkinElmer N612-1021 (OEM, instrument manufacturer′s quartz liner)
L × O.D. × I.D.
92 mm × 6.35 mm × 2.0 mm
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits PerkinElmer (AutoSystem)
Choose from one of the most recent versions:
Certificates of Analysis (COA)