MilliporeSigma
  • An electronic nose based on coated piezoelectric quartz crystals to certify ewes' cheese and to discriminate between cheese varieties.
An electronic nose based on coated piezoelectric quartz crystals to certify ewes' cheese and to discriminate between cheese varieties.
Sensors (Basel, Switzerland) (2012-03-23)
Vânia F Pais, João A B P Oliveira, Maria Teresa S R Gomes
ABSTRACT
An electronic nose based on coated piezoelectric quartz crystals was used to distinguish cheese made from ewes' milk, and to distinguish cheese varieties. Two sensors coated with Nafion and Carbowax could certify half the ewes' cheese samples, exclude 32 cheeses made from cow's milk and to classify half of the ewes' cheese samples as possibly authentic. Two other sensors, coated with polyvinylpyrrolidone and triethanolamine clearly distinguished between Flamengo, Brie, Gruyère and Mozzarella cheeses. Brie cheeses were further separated according to their origin, and Mozzarella grated cheese also appeared clearly separated from non-grated Mozzarella.
Sigma-Aldrich
2-Nonanone, ≥99%
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler
Sigma-Aldrich
Manganese(II) phthalocyanine
Supelco
Tenax® Porous Polymer Adsorbent, matrix Tenax TA, 80-100 mesh, bottle of 10 g