Skip to Content
MilliporeSigma
All Photos(2)

Documents

20896-U

Supelco

Tenax® TA Thermal Desorption Tube

greener alternative

glass, O.D. x I.D. x L 6 mm × 4 mm × 4.5 in., fritted, preconditioned, pkg of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

product name

Tenax® TA, glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 4.5 in., preconditioned, 35-60 mesh, pkg of 1 ea

material

glass TD tube

Quality Level

Agency

EPA TO-1,IP-1B

description

Sealed with TDS³ Storage Container

feature

fritted
preconditioned

packaging

pkg of 1 ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

active air sampling: suitable

O.D. × I.D. × L

6 mm × 4 mm × 4.5 in.

matrix

Tenax TA porous polymer

particle size

35-60 mesh

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with ACEM-9300, ACEM-900 850/890 TDU, MTDU, and OI Analytical Eclipse Air Tube Desorber

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

Each Tenax® TA sampling tube is made of glass. It is individually numbered, thermally conditioned, and batch tested for back-pressure and background. These prepacked Standard Sampling Tubes are sealed in exclusive storage containers. They incorporate a glass frit at the inlet, which increases their performance. They measure: O.D. × I.D. × L. 6 mm × 4 mm × 4.5 in.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. Thermal desorption tubes are reusable at least 50 to 100 times, and reduces the usage of accessories like transfer vials, caps, and pipette tips, thus aligns with “Waste Prevention”. Also, these tubes eliminate the usage of toxic solvents like CS2 to extract the VOCs of interest, and thus aligns with “Safer Solvents and Auxiliaries” principle. Click here for more information.

Legal Information

Tenax is a registered trademark of Buchem B.V.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service