Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04396

Supelco

Kieselguhr

calcined, purified

Synonym(s):

Infusorial earth

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116105
NACRES:
NA.21

grade

reagent grade

form

powder

quality

calcined, purified

technique(s)

LPLC: suitable

surface area

2-3.5 m2/g

impurities

≤1% HCl-soluble matter

loss

≤1% loss on ignition

matrix

Silica

matrix active group

silica

particle size

125 μm

pore size

170000 Å mean pore size

anion traces

chloride (Cl-): ≤200 mg/kg
sulfate (SO42-): ≤1000 mg/kg

cation traces

Fe: ≤500 mg/kg

separation technique

hydrophilic interaction (HILIC)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service