Skip to Content
MilliporeSigma
All Photos(17)

Key Documents

ZRB1703

Sigma-Aldrich

Anti-p-DNA-PKcs-Ser2056 Antibody, clone 1K3 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

DNA-dependent protein kinase catalytic subunit;EC:2.7.11.1;DNA-PK catalytic subunit;DNA-PKcs;DNPK1;p460

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1K3, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 469.09 kDa
observed mol wt ~350 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
peptide synthesis: suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PKRDC(5591)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1K3 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects DNA-dependent protein kinase catalytic subunit (DNA-PKcs) phosphorylated on serine 2056.

Immunogen

KLH-conjugated linear peptide corresponding to 14 amino acids surrounding phosphoserine 2056 in the internal region of human DNA-dependent protein kinase catalytic subunit (DNS-PKcs).

Application

Quality Control Testing

Evaluated by Immunohistochemistry (Paraffin) in human colon tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution of this antibody detected phospho-DNA-PKcs-Ser2056 in human colon tissue sections.

Tested applications

Peptide Inhibition Assay: Target band detection in lysates from Jurkat cells treated with 1 mM Camptothecin (3 h) was prevented by preblocking of a representative lot with the immunogen phosphopeptide, but not the corresponding non-phosphopeptide.

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected phospho-DNA-PKcs-Ser2056 in Jurkat cells treated with Campthothecin ( 1 mM; 3 h).

Affinity Binding Assay: A representative lot of this antibody bound phospho-DNA PKcs-Ser2056 with a KD of 9.3 x 10-7 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected phospho-DNA-PKcs-Ser2056 in Jurkat cells treated with Camptothecin.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

DNA-dependent protein kinase catalytic subunit (UniProt: P78527; also known as EC:2.7.11.1, DNA-PK catalytic subunit, DNA-PKcs, DNPK1, p460) is encoded by the PRKDC (also known as HYRC, HYRC1) gene (Gene ID: 5591) in human. DNA-PK is a heterotrimeric, nuclear serine/threonine-protein kinase that consists of PRKDC and the Ku p70/YRCC6-p86/XRCC5 dimer. It is involved in cell cycle control, DNA repair, and DNA damage responses. It serves as a molecular sensor for DNA damage and is involved in DNA non-homologous end joining required for double-strand break (DSB) repair and V(D)J recombination. It may also serve as a scaffold protein to aid the localization of DNA repair proteins to the site of damage. DNA-PK is also present at the ends of chromosomes, suggesting a role in the maintenance of telomeric stability and the prevention of chromosomal end fusion. Its catalytic activity is expressed upon binding to DNA. DNA-PKcs undergoes autophosphorylation at two clusters, the Thr2609 and the Ser2056, which are DNA damage-inducible phosphorylation sites. Autophosphorylation of the Thr2609 cluster is shown to be essential for hematopoietic development and protein synthesis in erythrocytes precursors. Autophosphorylation induces a conformational change that leads to remodeling of the DNA-PK complex, which is a requisite for efficient end processing and DNA repair. Although DNA-PKcs falls into the phosphatidylinositol (PI) 3-kinase family, it does not exhibit any no detectable activity toward lipids. Deficiency of DNA-PKcs is linked to severe combined immunodeficiency characterized by reduced or absent T and B cells, recurrent candidiasis, and lower respiratory tract infections. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Ma, Y., et al. (2004). Mol. Cell. 16(5); 701-713; Wechsler, T., et al. (2004). Proc. Nall. Acad. Sci. USA. 101(5); 1247-1252; Douglas, P., et al. (2002). Biochem. J. 368(1); 243-251).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service