Skip to Content
MilliporeSigma

08-110

ECL Cell Attachment Matrix

Synonym(s):

ECL Matrix

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NA.75
eCl@ss:
32160405
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

mouse

Quality Level

form

liquid

manufacturer/tradename

Upstate®

technique(s)

cell culture | mammalian: suitable

input

sample type neural stem cell(s)
sample type mesenchymal stem cell(s)
sample type induced pluripotent stem cell(s)
sample type hematopoietic stem cell(s)
sample type: human embryonic stem cell(s)
sample type epithelial cells
sample type pancreatic stem cell(s)

shipped in

dry ice

storage temp.

−20°C

General description

ECL Cell Attachment Matrix from Engelbreth-Holm-Swarm (EHS) mouse tumor.

Application

Cell culture attachment factor

Physical form

5 mg in 5 ml of 0.05 M Tris-HCl, pH 7.4, 0.15 M NaCl. Protein determined by the Bradford dye binding assay using gamma globulin as the standard. Frozen Solution.

Preparation Note

Stable for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing.

Analysis Note

Routinely evaluated in a Neurite Outgrowth Assay using PC12 rat pheochromocytoma cells.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B B Friday et al.
The Journal of cell biology, 149(3), 657-666 (2000-05-03)
Differentiation of skeletal muscle myoblasts follows an ordered sequence of events: commitment, cell cycle withdrawal, phenotypic differentiation, and finally cell fusion to form multinucleated myotubes. The molecular signaling pathways that regulate the progression are not well understood. Here we investigate
Extracellular matrix regulates Sertoli cell differentiation, testicular cord formation, and germ cell development in vitro.
Hadley, M A, et al.
The Journal of cell biology, 101, 1511-1522 (1985)
Alteration of Ca2+ dependence of neurotransmitter release by disruption of Ca2+ channel/syntaxin interaction
Rettig, J., et al
The Journal of Neuroscience, 17, 6647-6656 (1997)
Basement membrane complexes with biological activity.
Kleinman, H K, et al.
Biochemistry, 25, 312-318 (1986)
F B Gertler et al.
Cell, 87(2), 227-239 (1996-10-18)
Drosophila Enabled is required for proper formation of axonal structures and is genetically implicated in signaling pathways mediated by Drosophila AbI. We have identified two murine proteins, Mena and Evl, that are highly related to Enabled as well as VASP

Related Content

As the focus of stem cell research undergoes a transition from animal to human models, researchers are in critical need of validated products to support the isolation, maintenance, differentiation, and characterization of human stem cells. While many reagents designed for rodent systems can be applied to human stem cell studies, they are not truly optimized for robust human stem cell culture or analysis. This is why human stem cell researchers have always trusted EMD Millipore, the first provider of commercially available human embryonic stem cells and human neural stem cell lines, to accelerate their research. All of our human stem cell systems are extensively tested in defined media culture, and differentiated progeny are comprehensively characterized with highly validated antibodies and detection reagents.

Global Trade Item Number

SKUGTIN
08-11004053252677632

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service