Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

CLS354234

Corning® Matrigel® Basement Membrane Matrix

pack of 10 mL

Synonym(s):

Basement membrane matrix, Cell culture matrix, Matrigel matrix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116155
NACRES:
NA.7

packaging

pack of 10 mL

manufacturer/tradename

Corning 354234

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.

Quality

Mouse colonies routinely screened for pathogens via Mouse Antibody Production (MAP) testing
Extensive PCR testing is performed to screen for a number of pathogensTested and found negative for bacteria, fungi, and mycoplasma
Protein concentrations determined by Lowry method
Endotoxin units measured by Limulus Amoebocyte Lysate assay
Matrigel Matrix gel stability tested for a period of 14 days at 37°C
Biological activity determined for each lot using a neurite outgrowth assay.
Source
Engelbreth-Holm-Swarm Mouse Tumor

Preparation and Storage
Store at -20°C. Avoid multiple freeze-thaws.

Application

Cell Growth and Differentiation
Metabolism/Toxicology Studies
Invasion Assays
in vitro and in vivo Angiogenesis Assays
in vivo Angiogenesis Studies and Augmentation of Tumors in Immunosuppressed Mice


For Customers in California only: WARNING Please refer to Safety Data Sheet (SDS) Section 15 or Letter in Lieu for more information regarding California Proposition 65.

Legal Information

Corning is a registered trademark of Corning, Inc.
Matrigel is a registered trademark of Corning, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Learn how the Corning® Matrigel® Basement Membrane Matrix can be used to boost your 3D Cell Culture activities.

Learn how the Corning® Matrigel® Basement Membrane Matrix can be used to boost your 3D Cell Culture activities.

Learn how the Corning® Matrigel® Basement Membrane Matrix can be used to boost your 3D Cell Culture activities.

Learn the results that come from changing the protein concentration of Corning® Matrigel®.

See All

Related Content

Culture in 2.5D using small intestine duodenum organoids and Millicell® hanging cell culture inserts to monitor long-term drug side effects and toxicity.

Culture in 2.5D using small intestine duodenum organoids and Millicell® hanging cell culture inserts to monitor long-term drug side effects and toxicity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service