Skip to Content
MilliporeSigma
All Photos(2)

Documents

Z764655

MAGic Clamp tube rack

to hold, 15 x 50 mL tubes

Synonym(s):

tube clamps

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122413
NACRES:
NB.12

feature

to hold 15 x 50 mL tubes

packaging

pkg of 1 ea

manufacturer/tradename

Benchmark Scientific H1000-MR-T50

Looking for similar products? Visit Product Comparison Guide

General description

  • Extremely compact
  • Exceptional temperature uniformity
  • Horizontally circular 19 mm orbit for aeration and mixing

Legal Information

MAGic Clamp is a trademark of Benchmark Scientific, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service